Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP39
Entrez Gene ID
839314
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT1G04080, F20D22.14
Chromosome Location
Chr.1: 1051312 - 1056827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160830.2 NP_001154302.1 6 - 7 1415 85
NM_001197976.1 NP_001184905.1 11 - 12 1934 85
NM_001331460.1 NP_001322270.1 8 - 9 1824 85
NM_001331461.1 NP_001322271.1 6 - 7 1235 85
NM_100289.4 NP_563700.1 9 - 10 1738 85
GenBank mRNA AY056321.1 - 9 - 10 1703 85
AY087878.1 - 9 - 10 1705 85
AY091187.1 - 8 - 9 1555 85

View Full Product Details