Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIA
Entrez Gene ID
839480
Gene Name
myosin XI A
Gene Aliases
AT1G04600, ATXIA, T1G11.15, T1G11_15, myosin XI A
Chromosome Location
Chr.1: 1261762 - 1272588 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100339.2 NP_171954.1 2 - 3 489 127

View Full Product Details