Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTF18
Entrez Gene ID
839431
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT1G04730, CHROMOSOME TRANSMISSION FIDELITY 18, T1G11.3, T1G11_3
Chromosome Location
Chr.1: 1325099 - 1331169 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331495.1 NP_001322020.1 21 - 22 2820 91
NM_001331496.1 NP_001322021.1 19 - 20 2292 91
NM_001331497.1 NP_001322022.1 21 - 22 2843 91
NM_100352.3 NP_171966.2 21 - 22 2860 91

View Full Product Details