Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EFE
- Entrez Gene ID
-
839345
- Gene Name
- ethylene-forming enzyme
- Gene Aliases
-
1-AMINOCYCLOPROPANE-1-CARBOXYLATE OXIDASE, ACO4, AT1G05010, EAT1, ETHYLENE-FORMING ENZYME, T7A14.12, T7A14_12, ethylene forming enzyme, ethylene-forming enzyme
- Chromosome Location
-
Chr.1: 1431177 - 1433010 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100380.4 |
NP_171994.1 |
2 - 3 |
642 |
77 |
GenBank mRNA |
AY150384.1 |
- |
2 - 3 |
327 |
77 |
View Full Product Details