Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT75B2
Entrez Gene ID
837055
Gene Name
UDP-glucosyl transferase 75B2
Gene Aliases
AT1G05530, T25N20.18, UDP-GLUCOSYL TRANSFERASE 2, UDP-glucosyl transferase 75B2, UGT2
Chromosome Location
Chr.1: 1636249 - 1637914 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100432.2 NP_172044.1 1 - 1 819 137
GenBank mRNA KJ139029.1 - 1 - 1 768 137

View Full Product Details