Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APS2
- Entrez Gene ID
-
837066
- Gene Name
- ADP-glucose pyrophosphorylase small subunit 2
- Gene Aliases
-
ADP-GLUCOSE PYROPHOSPHORYLASE, ADP-glucose pyrophosphorylase small subunit 2, AT1G05610, F3F20.6, F3F20_6
- Chromosome Location
-
Chr.1: 1673861 - 1676077 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100441.3 |
NP_172052.2 |
4 - 5 |
1115 |
92 |
View Full Product Details