Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMB1444
Entrez Gene ID
6241227
Gene Name
basic helix loop helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT1G06150, AtRLP53, EMBRYO DEFECTIVE 1444, LHL1, LONESOME HIGHWAY LIKE 1, T21E18.20, T21E18_20, receptor like protein 53
Chromosome Location
Chr.1: 1868989 - 1873730 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197992.2 NP_001184921.1 9 - 10 2530 74
NM_001331616.1 NP_001318931.1 9 - 10 2536 74

View Full Product Details