Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT6
Entrez Gene ID
837189
Gene Name
galacturonosyltransferase 6
Gene Aliases
AT1G06780, F4H5.13, F4H5_13, galacturonosyltransferase 6
Chromosome Location
Chr.1: 2083311 - 2087141 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197996.1 NP_001184925.1 8 - 9 1914 89
NM_100555.3 NP_563771.1 8 - 9 1830 89
GenBank mRNA AY045900.1 - 8 - 9 1762 89
AY091452.1 - 8 - 9 1497 89
KJ139026.1 - 8 - 9 1497 89

View Full Product Details