Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX14
Entrez Gene ID
837207
Gene Name
cation/hydrogen exchanger 14
Gene Aliases
AT1G06970, ATCHX14, F10K1.31, F10K1_31, cation/hydrogen exchanger 14
Chromosome Location
Chr.1: 2137871 - 2140975 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100570.2 NP_172178.1 2 - 3 1489 94
GenBank mRNA DQ446234.1 - 2 - 3 1251 94
EF571900.1 - 2 - 3 1251 94

View Full Product Details