Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NEK5
- Entrez Gene ID
-
821634
- Gene Name
- Serine/Threonine kinase catalytic domain protein
- Gene Aliases
-
AT3G20860, ATNEK5, MOE17.17, NIMA-RELATED KINASE5, NIMA-related kinase 5
- Chromosome Location
-
Chr.3: 7306180 - 7308434 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112977.2 |
NP_188722.2 |
3 - 4 |
220 |
124 |
View Full Product Details