Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFU4
Entrez Gene ID
821647
Gene Name
NFU domain protein 4
Gene Aliases
ARABIDOPSIS THALIANA NFU DOMAIN PROTEIN 2, AT3G20970, ATNFU2, NFU domain protein 4
Chromosome Location
Chr.3: 7348157 - 7350341 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035658.1 NP_001030735.1 5 - 6 855 78
NM_112989.3 NP_566673.1 5 - 6 897 78
GenBank mRNA AF370537.1 - 5 - 6 857 78
AJ512936.1 - 5 - 6 803 78
AY072509.1 - 5 - 6 777 78

View Full Product Details