Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCD7
Entrez Gene ID
819107
Gene Name
carotenoid cleavage dioxygenase 7
Gene Aliases
AT2G44990, ATCCD7, MAX3, T14P1.21, carotenoid cleavage dioxygenase 7
Chromosome Location
Chr.2: 18558885 - 18561621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337111.1 NP_001318427.1 1 - 2 758 88
NM_001337112.1 NP_001324720.1 1 - 2 788 88
NM_130064.5 NP_182026.5 1 - 2 811 88

View Full Product Details