Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD51C
Entrez Gene ID
819136
Gene Name
RAS associated with diabetes protein 51C
Gene Aliases
AT2G45280, ATRAD51C, F4L23.21, RAS associated with diabetes protein 51C
Chromosome Location
Chr.2: 18670009 - 18672469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202820.1 NP_001189749.1 6 - 7 880 98
NM_130091.2 NP_566040.1 6 - 7 808 98
GenBank mRNA AB062456.1 - 6 - 7 808 98

View Full Product Details