Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD19
Entrez Gene ID
819149
Gene Name
LOB domain-containing protein 19
Gene Aliases
AT2G45410, F4L23.8, LOB domain-containing protein 19
Chromosome Location
Chr.2: 18712220 - 18713211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130103.3 NP_182065.1 1 - 2 413 73
GenBank mRNA AB473856.1 - 1 - 2 313 73

View Full Product Details