Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP76C4
- Entrez Gene ID
-
819163
- Gene Name
- cytochrome P450, family 76, subfamily C, polypeptide 4
- Gene Aliases
-
''cytochrome P450, AT2G45550, F17K2.8, cytochrome P450, family 76, polypeptide 4, polypeptide 4'', subfamily C
- Chromosome Location
-
Chr.2: 18773517 - 18775797 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130117.2 |
NP_182079.1 |
1 - 2 |
536 |
77 |
View Full Product Details