Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP76C3
Entrez Gene ID
819166
Gene Name
cytochrome P450, family 76, subfamily C, polypeptide 3
Gene Aliases
''cytochrome P450, AT2G45580, F17K2.11, cytochrome P450, family 76, polypeptide 3, polypeptide 3'', subfamily C
Chromosome Location
Chr.2: 18782231 - 18784530 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337145.1 NP_001323695.1 1 - 2 452 114
NM_130120.4 NP_182082.2 1 - 2 635 114

View Full Product Details