Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRM
Entrez Gene ID
819210
Gene Name
transcription regulatory protein SNF2
Gene Aliases
ARABIDOPSIS THALIANA BRAHMA, AT2G46020, ATBRM, BRAHMA, CHA2, CHR2, CHROMATIN REMODELING 2, T3F17.33
Chromosome Location
Chr.2: 18922533 - 18932055 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337166.1 NP_001318432.1 14 - 15 6518 134
NM_001337167.1 NP_001323762.1 14 - 15 6650 134
NM_001337168.1 NP_001323760.1 14 - 15 6495 134
NM_001337169.1 NP_001323761.1 14 - 15 6621 134
NM_130165.3 NP_182126.2 13 - 14 5952 134
NM_201966.2 NP_973695.1 13 - 14 5955 134
GenBank mRNA AJ703891.1 - 13 - 14 5955 134

View Full Product Details