Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KPHMT1
Entrez Gene ID
819219
Gene Name
ketopantoate hydroxymethyltransferase 1
Gene Aliases
AT2G46110, PANB1, T3F17.24, ketopantoate hydroxymethyltransferase 1
Chromosome Location
Chr.2: 18954570 - 18956582 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130174.5 NP_182135.1 1 - 2 571 64
GenBank mRNA AK227849.1 - 1 - 2 519 64
BT004820.1 - 1 - 2 400 64

View Full Product Details