Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRF5
Entrez Gene ID
819239
Gene Name
cytokinin response factor 5
Gene Aliases
AT2G46310, T3F17.4, cytokinin response factor 5
Chromosome Location
Chr.2: 19011462 - 19013018 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130194.5 NP_182154.1 1 - 1 1306 171
GenBank mRNA AK227661.1 - 1 - 1 1265 171

View Full Product Details