Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC6
Entrez Gene ID
819269
Gene Name
laccase 6
Gene Aliases
AT2G46570, F13A10.10, laccase 6
Chromosome Location
Chr.2: 19126272 - 19129069 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130222.2 NP_182180.1 1 - 2 713 65

View Full Product Details