Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPa3
Entrez Gene ID
819299
Gene Name
pyrophosphorylase 3
Gene Aliases
AT2G46860, AtPPa3, F19D11.23, F19D11_23, pyrophosphorylase 3
Chromosome Location
Chr.2: 19253670 - 19255328 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130253.3 NP_182209.1 6 - 7 589 68
GenBank mRNA BT025278.1 - 5 - 6 529 68

View Full Product Details