Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCC4
- Entrez Gene ID
-
819392
- Gene Name
- multidrug resistance-associated protein 4
- Gene Aliases
-
AT2G47800, ATMRP4, ATP-binding cassette C4, EST3, F17A22.19, MRP4, multidrug resistance-associated protein 4
- Chromosome Location
-
Chr.2: 19574666 - 19580615 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130347.5 |
NP_182301.1 |
6 - 7 |
3898 |
91 |
GenBank mRNA |
AF243509.1 |
- |
6 - 7 |
3620 |
91 |
View Full Product Details