Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- REF4
- Entrez Gene ID
-
819423
- Gene Name
- reduced epidermal fluorescence 4
- Gene Aliases
-
AT2G48110, F11L15.1, MED33B, REDUCED EPIDERMAL FLUORESCENCE 4, T9J23.25
- Chromosome Location
-
Chr.2: 19673293 - 19679711 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130378.5 |
NP_566125.4 |
3 - 4 |
809 |
65 |
View Full Product Details