Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAC
Entrez Gene ID
819424
Gene Name
pale cress protein (PAC)
Gene Aliases
AT2G48120, F11L15.2, PALE CRESS
Chromosome Location
Chr.2: 19679730 - 19681546 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084610.1 NP_001078079.1 6 - 7 817 71
NM_130379.2 NP_182332.1 6 - 7 835 71
GenBank mRNA AK117768.1 - 6 - 7 745 71
AY086817.1 - 6 - 7 772 71
BT005304.1 - 6 - 7 695 71

View Full Product Details