Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMPK13
Entrez Gene ID
837303
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G07880, F24B9.3, F24B9_3
Chromosome Location
Chr.1: 2433896 - 2435858 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035913.2 NP_001030990.1 2 - 3 416 80
NM_100662.4 NP_172266.2 2 - 3 416 80
GenBank mRNA BT015822.1 - 2 - 3 303 80

View Full Product Details