Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATCHX5
- Entrez Gene ID
-
837336
- Gene Name
- Cation/hydrogen exchanger family protein
- Gene Aliases
-
AT1G08150, CATION/H+ EXCHANGER 5, CHX5, T6D22.24
- Chromosome Location
-
Chr.1: 2556191 - 2559228 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100690.3 |
NP_172294.2 |
1 - 2 |
462 |
71 |
View Full Product Details