Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGSIP5
- Entrez Gene ID
-
837420
- Gene Name
- plant glycogenin-like starch initiation protein 5
- Gene Aliases
-
AT1G08990, F7G19.14, F7G19_14, GUX5, Glucuronic Acid Substitution of Xylan 5, plant glycogenin-like starch initiation protein 5
- Chromosome Location
-
Chr.1: 2888427 - 2890965 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100770.5 |
NP_172373.3 |
2 - 3 |
1536 |
111 |
GenBank mRNA |
KJ139018.1 |
- |
2 - 3 |
1210 |
111 |
View Full Product Details