Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-B15
Entrez Gene ID
837435
Gene Name
phloem protein 2-B15
Gene Aliases
AT1G09155, AtPP2-B15, phloem protein 2-B15
Chromosome Location
Chr.1: 2949621 - 2950989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100785.3 NP_563837.1 2 - 3 515 104

View Full Product Details