Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP-PRT2
Entrez Gene ID
837509
Gene Name
ATP phosphoribosyl transferase 2
Gene Aliases
AT1G09795, ATATP-PRT2, ATP PHOSPHORIBOSYL TRANSFERASE 2, ATP phosphoribosyl transferase 2, HISN1B
Chromosome Location
Chr.1: 3173534 - 3176910 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100852.2 NP_563853.1 7 - 8 884 101
GenBank mRNA AK118503.1 - 7 - 8 870 101
BT002054.1 - 7 - 8 870 101
BT008501.1 - 7 - 8 830 101

View Full Product Details