Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFA1F
Entrez Gene ID
837606
Gene Name
ADP-ribosylation factor A1F
Gene Aliases
ADP-ribosylation factor A1F, AT1G10630, ATARFA1F, F20B24.7, F20B24_7
Chromosome Location
Chr.1: 3512739 - 3514824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331919.1 NP_001321560.1 4 - 5 669 68
NM_100939.4 NP_172533.2 4 - 5 538 68
GenBank mRNA AK226308.1 - 4 - 5 439 68
BT014776.1 - 4 - 5 388 68
BT015004.1 - 3 - 4 340 68

View Full Product Details