Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD4
Entrez Gene ID
837629
Gene Name
ARF-GAP domain 4
Gene Aliases
ARF-GAP domain 4, AT1G10870, T19D16.20, T19D16_20
Chromosome Location
Chr.1: 3616610 - 3623806 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331933.1 NP_001320312.1 1 - 2 242 138
NM_100962.4 NP_172556.2 1 - 2 242 138
GenBank mRNA AJ130878.1 - 1 - 2 177 138

View Full Product Details