Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUB
Entrez Gene ID
837654
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT1G11130, AT1G11140, SCM, SCRAMBLED, SRF9, STRUBBELIG, STRUBBELIG-RECEPTOR FAMILY 9, T19D16.8
Chromosome Location
Chr.1: 3722708 - 3727450 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331960.1 NP_001321706.1 2 - 3 233 83
NM_100986.4 NP_172580.2 3 - 4 717 83
GenBank mRNA AF399923.1 - 3 - 4 506 83
AK227100.1 - 3 - 4 518 83
AY091019.1 - 3 - 4 568 83
AY117318.1 - 3 - 4 290 83
FJ708632.1 - 3 - 4 290 83

View Full Product Details