Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BFN1
Entrez Gene ID
837660
Gene Name
bifunctional nuclease i
Gene Aliases
AT1G11190, ENDO1, ENDONUCLEASE 1, T28P6.14, T28P6_14, bifunctional nuclease i
Chromosome Location
Chr.1: 3750066 - 3752780 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100991.2 NP_172585.1 1 - 2 206 70
GenBank mRNA AY040016.1 - 1 - 2 206 70
AY079368.1 - 1 - 2 122 70
AY088393.1 - 1 - 2 206 70

View Full Product Details