Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP8
Entrez Gene ID
835717
Gene Name
actin-related protein 8
Gene Aliases
AT5G56180, AT5G56185, ATARP8, MDA7.26, actin-related protein 8
Chromosome Location
Chr.5: 22737325 - 22741100 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345181.1 NP_001330467.1 8 - 9 1321 112
NM_125000.4 NP_568836.2 9 - 10 1200 112
GenBank mRNA AF507916.1 - 9 - 10 1093 112

View Full Product Details