Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MKK6
- Entrez Gene ID
-
835759
- Gene Name
- MAP kinase kinase 6
- Gene Aliases
-
ANQ1, ARABIDOPSIS NQK1, ARABIDOPSIS THALIANA MAP KINASE KINASE 6, AT5G56580, ATMKK6, MAP kinase kinase 6, MIK19.2, MIK19_2
- Chromosome Location
-
Chr.5: 22904622 - 22906764 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125041.3 |
NP_200469.1 |
5 - 6 |
853 |
73 |
GenBank mRNA |
AB104460.1 |
- |
5 - 6 |
750 |
73 |
View Full Product Details