Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK7
Entrez Gene ID
835764
Gene Name
phosphofructokinase 7
Gene Aliases
AT5G56630, MIK19.8, MIK19_8, phosphofructokinase 7
Chromosome Location
Chr.5: 22923804 - 22926963 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125046.3 NP_568842.1 6 - 7 1108 75
GenBank mRNA AF360207.1 - 6 - 7 990 75
AY040055.1 - 6 - 7 601 75

View Full Product Details