Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CKX3
Entrez Gene ID
835799
Gene Name
cytokinin oxidase 3
Gene Aliases
AT5G56970, ATCKX3, CYTOKININ OXIDASE, MHM17.8, MHM17_8, cytokinin oxidase 3
Chromosome Location
Chr.5: 23044839 - 23048292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345226.1 NP_001331716.1 3 - 4 1036 121
NM_125079.3 NP_200507.1 3 - 4 1044 121
GenBank mRNA AF303979.1 - 3 - 4 997 121
BT033021.1 - 3 - 4 997 121

View Full Product Details