Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIR1
Entrez Gene ID
835813
Gene Name
Auxin efflux carrier family protein
Gene Aliases
AGR, AGR1, AGRAVITROPIC ROOT, AGRAVITROPIC ROOT 1, ARABIDOPSIS THALIANA PIN-FORMED 2, AT5G57090, ATPIN2, AUXIN TRANSPORT PROTEIN EIR1, ETHYLENE INSENSITIVE ROOT 1, MM31, MUL3.3, MUL3_3, PIN-FORMED 2, PIN2, POLAR-AUXIN-TRANSPORT EFFLUX COMPONENT AGRAVITROPIC 1, WAV6, WAVY ROOTS 6
Chromosome Location
Chr.5: 23100516 - 23105228 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125091.4 NP_568848.1 1 - 2 524 64
GenBank mRNA AF056026.1 - 1 - 2 293 64
AF086906.1 - 1 - 2 348 64
AF093241.1 - 1 - 2 298 64
AK176358.1 - 1 - 2 331 64
AY078965.1 - 1 - 2 327 64
BT001152.1 - 1 - 2 275 64

View Full Product Details