Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT5G57300
Entrez Gene ID
835835
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT5G57300, MJB24.11, MJB24_11
Chromosome Location
Chr.5: 23208463 - 23211139 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085291.3 NP_001078760.1 5 - 6 607 123
NM_001203629.1 NP_001190558.1 5 - 6 600 123
NM_001345255.1 NP_001332423.1 4 - 5 479 123
NM_125112.5 NP_200540.1 5 - 6 557 123
GenBank mRNA AY050939.1 - 5 - 6 556 123
AY091173.1 - 4 - 5 430 123

View Full Product Details