Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCH4
Entrez Gene ID
835860
Gene Name
Xyloglucan endotransglucosylase/hydrolase family protein
Gene Aliases
AT5G57560, MUA2.13, MUA2_13, Touch 4, XTH22, XYLOGLUCAN ENDOTRANSGLYCOSYLASE, xyloglucan endotransglucosylase/hydrolase 22
Chromosome Location
Chr.5: 23307006 - 23308680 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125137.4 NP_200564.1 1 - 2 710 66
GenBank mRNA AF051338.1 - 1 - 2 265 66
AF367262.1 - 1 - 2 311 66
AF446881.1 - 1 - 2 265 66
AY052712.1 - 1 - 2 311 66
AY055102.1 - 1 - 2 265 66

View Full Product Details