Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPS1
Entrez Gene ID
835898
Gene Name
multipolar spindle 1
Gene Aliases
ARABIDOPSIS THALIANA PUTATIVE RECOMBINATION INITIATION DEFECTS 2, AT5G57880, ATPRD2, MULTIPOLAR SPINDLE 1, PRD2, PUTATIVE RECOMBINATION INITIATION DEFECTS 2
Chromosome Location
Chr.5: 23443962 - 23447244 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345277.1 NP_001318822.1 7 - 8 689 125

View Full Product Details