Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPB5C
- Entrez Gene ID
-
835909
- Gene Name
- RNA polymerase II fifth largest subunit, C
- Gene Aliases
-
''RNA polymerase II fifth largest subunit, AT5G57980, C, C'', MTI20.24, MTI20_24, RNA polymerase II fifth largest subunit
- Chromosome Location
-
Chr.5: 23468959 - 23469856 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125183.2 |
NP_200606.1 |
1 - 2 |
297 |
72 |
View Full Product Details