Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YKT61
Entrez Gene ID
835918
Gene Name
SNARE-like superfamily protein
Gene Aliases
AT5G58060, ATGP1, ATYKT61, K21L19.5, K21L19_5
Chromosome Location
Chr.5: 23498139 - 23500384 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161313.1 NP_001154785.1 6 - 7 749 95
NM_125191.4 NP_200614.1 5 - 6 683 95
GenBank mRNA AF325040.2 - 5 - 6 675 95
AF385703.1 - 5 - 6 675 95
U64918.1 - 5 - 6 612 95

View Full Product Details