Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RR18
Entrez Gene ID
835920
Gene Name
response regulator 18
Gene Aliases
ARR18, AT5G58080, K21L19.7, K21L19_7, response regulator 18
Chromosome Location
Chr.5: 23501605 - 23504169 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125193.4 NP_200616.4 4 - 5 745 83

View Full Product Details