Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTL
Entrez Gene ID
835934
Gene Name
transthyretin-like protein
Gene Aliases
ALNS, AT5G58220, MCK7.9, MCK7_9, allantoin synthase, transthyretin-like protein
Chromosome Location
Chr.5: 23554386 - 23556001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037016.2 NP_001032093.1 1 - 2 301 101
NM_001037017.3 NP_001032094.1 1 - 2 421 101
NM_125207.5 NP_200630.1 1 - 2 421 101
GenBank mRNA AK226943.1 - 1 - 2 301 101
AY062771.1 - 1 - 2 299 101
AY081647.1 - 1 - 2 281 101

View Full Product Details