Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTL
Entrez Gene ID
835934
Gene Name
transthyretin-like protein
Gene Aliases
ALNS, AT5G58220, MCK7.9, MCK7_9, allantoin synthase, transthyretin-like protein
Chromosome Location
Chr.5: 23554386 - 23556001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125207.5 NP_200630.1 3 - 4 679 72
GenBank mRNA AY062771.1 - 3 - 4 557 72
AY081647.1 - 3 - 4 539 72

View Full Product Details