Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB25
Entrez Gene ID
835939
Gene Name
ABC transporter of the mitochondrion 3
Gene Aliases
ABC transporter of the mitochondrion 3, ARABIDOPSIS THALIANA ABC TRANSPORTER OF THE MITOCHONDRION 3, AT5G58270, ATATM3, ATM3, ATP-binding cassette B25, HALF-MOLECULE ABC TRANSPORTER ATM3, MCK7.14, MCK7_14, STA1, STARIK 1
Chromosome Location
Chr.5: 23561890 - 23567244 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125212.3 NP_200635.1 1 - 2 924 64
GenBank mRNA AF287699.1 - 1 - 2 646 64
AF360334.1 - 1 - 2 669 64
AY142686.1 - 1 - 2 646 64

View Full Product Details