Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPT3
Entrez Gene ID
835941
Gene Name
regulatory particle triple-A ATPase 3
Gene Aliases
26S PROTEASOME AAA-ATPASE SUBUNIT RPT3, AT5G58290, MCK7.16, MCK7_16, regulatory particle triple-A ATPase 3
Chromosome Location
Chr.5: 23569052 - 23571854 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125214.6 NP_200637.1 1 - 2 566 90
GenBank mRNA AF123392.1 - 1 - 2 504 90
AY070466.1 - - 979 90
BT020373.1 - 1 - 2 463 90

View Full Product Details