Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPT1
Entrez Gene ID
836020
Gene Name
copper transporter 1
Gene Aliases
AT5G59030, K19M22.24, copper transporter 1
Chromosome Location
Chr.5: 23833493 - 23834745 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125293.4 NP_200711.1 1 - 1 963 94

View Full Product Details