Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPT3
Entrez Gene ID
836021
Gene Name
copper transporter 3
Gene Aliases
AT5G59040, K18B18.3, K18B18_3, copper transporter 3
Chromosome Location
Chr.5: 23836625 - 23837350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125294.3 NP_200712.1 1 - 1 509 79
GenBank mRNA DQ447093.1 - 1 - 1 410 79

View Full Product Details